Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615474.3 | XP_005615531.1 | 1421 | 90 | ||
XM_005615479.3 | XP_005615536.1 | 1252 | 90 | |||
XM_014112545.2 | XP_013968020.1 | 1756 | 90 | |||
XM_014112555.2 | XP_013968030.1 | 1794 | 90 | |||
XM_014112558.2 | XP_013968033.1 | 1826 | 90 | |||
XM_014112566.2 | XP_013968041.1 | 1788 | 90 | |||
XM_014112572.2 | XP_013968047.1 | 1770 | 90 | |||
XM_014112577.2 | XP_013968052.1 | 1682 | 90 | |||
XM_022417720.1 | XP_022273428.1 | 1722 | 90 | |||
XM_022417724.1 | XP_022273432.1 | 1697 | 90 | |||
XM_022417725.1 | XP_022273433.1 | 1620 | 90 | |||
XM_022417731.1 | XP_022273439.1 | 1754 | 90 | |||
XM_022417736.1 | XP_022273444.1 | 1736 | 90 | |||
XM_022417744.1 | XP_022273452.1 | 1612 | 90 | |||
XM_022417747.1 | XP_022273455.1 | 1726 | 90 | |||
XM_022417748.1 | XP_022273456.1 | 1535 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris |