Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639141.3 | XP_003639189.1 | 355 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris |