Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114615.2 | XP_013970090.1 | 2 | 349 | 82 | |
XM_022420063.1 | XP_022275771.1 | 2 | 257 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |