Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022409695.1 | XP_022265403.1 | 3 - 4 | 812 | 150 | |
XM_843990.5 | XP_849083.1 | 3 - 4 | 836 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |