Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENDOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846067.4 | XP_851160.3 | 2 | 738 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried Amplicon spans exons and probe does not span exons |