Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849349.5 | XP_854442.1 | 1 - 2 | 413 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons |