Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014121011.2 | XP_013976486.1 | 1 - 2 | 376 | 104 | |
XM_533233.6 | XP_533233.2 | 2 - 3 | 312 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |