Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637720.3 | XP_005637777.1 | 4 - 5 | 1410 | 148 | |
XM_022411763.1 | XP_022267471.1 | 3 - 4 | 1166 | 148 | ||
XM_535013.6 | XP_535013.2 | 4 - 5 | 1304 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |