Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_541947.3 | XP_541947.1 | 3 | 717 | 178 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |