Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022425639.1 | XP_022281347.1 | 6 | 1078 | 159 | |
XM_849850.6 | XP_854943.3 | 6 | 1170 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |