Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK38L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284490.1 | NP_001271419.1 | 3 - 4 | 224 | 136 | |
GenBank mRNA | GU811153.1 | - | 3 - 4 | 226 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |