Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK38L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284490.1 | NP_001271419.1 | 5 - 6 | 437 | 172 | |
GenBank mRNA | GU811153.1 | - | 5 - 6 | 439 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |