Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635064.3 | XP_005635121.1 | 3 | 790 | 185 | |
XM_005635065.2 | XP_005635122.1 | 3 | 889 | 185 | ||
XM_005635066.3 | XP_005635123.1 | 3 | 847 | 185 | ||
XM_005635068.3 | XP_005635125.1 | 2 | 710 | 185 | ||
XM_005635069.2 | XP_005635126.1 | 2 | 771 | 185 | ||
XM_022409122.1 | XP_022264830.1 | 3 | 639 | 185 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |