Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638537.3 | XP_005638594.1 | 2 | 1330 | 91 | |
XM_005638538.3 | XP_005638595.1 | 2 | 1456 | 91 | ||
XM_022412572.1 | XP_022268280.1 | 2 | 1212 | 91 | ||
XM_535526.6 | XP_535526.3 | 2 | 1570 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |