Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634012.3 | XP_005634069.1 | 2 | 3107 | 81 | |
XM_005634013.3 | XP_005634070.1 | 2 | 3479 | 81 | ||
XM_844279.4 | XP_849372.1 | 2 | 3067 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |