Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4K2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_543953.6 | XP_543953.2 | 3 - 4 | 858 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |