Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544602.6 | XP_544602.2 | 2 | 1163 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |