Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629230.3 | XP_005629287.1 | 3 | 884 | 140 | |
XM_005629231.3 | XP_005629288.1 | 2 | 762 | 140 | ||
XM_005629232.3 | XP_005629289.1 | 2 | 768 | 140 | ||
XM_022403323.1 | XP_022259031.1 | 4 | 930 | 140 | ||
XM_022403324.1 | XP_022259032.1 | 3 | 814 | 140 | ||
XM_022403325.1 | XP_022259033.1 | 2 | 768 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |