Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630500.3 | XP_005630557.1 | 3 | 652 | 139 | |
XM_005630501.3 | XP_005630558.1 | 3 | 651 | 139 | ||
XM_022404762.1 | XP_022260470.1 | 3 | 582 | 139 | ||
XM_540206.6 | XP_540206.3 | 3 | 652 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |