Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636818.3 | XP_005636875.1 | 3 - 4 | 654 | 136 | |
XM_005636819.2 | XP_005636876.1 | 3 - 4 | 2377 | 136 | ||
XM_022411157.1 | XP_022266865.1 | 2 - 3 | 2008 | 136 | ||
XM_845163.4 | XP_850256.1 | 3 - 4 | 623 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |