Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022410959.1 | XP_022266667.1 | 3 | 636 | 62 | |
XM_022410960.1 | XP_022266668.1 | 3 | 498 | 62 | ||
XM_849304.5 | XP_854397.1 | 3 | 617 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |