Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_847473.4 | XP_852566.1 | 3 - 4 | 413 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |