Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_546803.5 | XP_546803.1 | 3 | 1359 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |