Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROKR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022409240.1 | XP_022264948.1 | 2 | 2283 | 146 | |
XM_845933.4 | XP_851026.1 | 3 | 1381 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |