Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022422914.1 | XP_022278622.1 | 353 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris |