Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX6-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_844065.3 | XP_849158.1 | 2888 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |