Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD9L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_539422.6 | XP_539422.3 | 2 | 387 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |