Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZADH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615306.1 | XP_005615363.1 | 1085 | 151 | ||
XM_014121375.2 | XP_013976850.1 | 749 | 151 | |||
XM_022410465.1 | XP_022266173.1 | 753 | 151 | |||
XM_022410498.1 | XP_022266206.1 | 755 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |