Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634768.3 | XP_005634825.1 | 3 | 1789 | 77 | |
XM_852855.5 | XP_857948.1 | 3 | 1238 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |