Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014121652.2 | XP_013977127.1 | 1458 | 140 | ||
GenBank mRNA | DQ202369.1 | - | 1282 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |