Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433755.4 | XP_003433803.2 | 433 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |