Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629093.3 | XP_005629150.1 | 6 - 7 | 969 | 99 | |
XM_014119480.2 | XP_013974955.1 | 6 - 7 | 710 | 99 | ||
XM_014119481.2 | XP_013974956.1 | 5 - 6 | 620 | 99 | ||
XM_014119482.2 | XP_013974957.1 | 5 - 6 | 431 | 99 | ||
XM_014119483.2 | XP_013974958.1 | 4 - 5 | 359 | 99 | ||
XM_022428038.1 | XP_022283746.1 | 6 - 7 | 969 | 99 | ||
XM_022428040.1 | XP_022283748.1 | 4 - 5 | 612 | 99 | ||
XM_022428042.1 | XP_022283750.1 | 4 - 5 | 467 | 99 | ||
XM_845756.4 | XP_850849.2 | 6 - 7 | 966 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |