Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536362.5 | XP_536362.3 | 1 - 2 | 233 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |