Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625749.2 | XP_005625806.1 | 2 - 3 | 638 | 141 | |
XM_022424288.1 | XP_022279996.1 | 2 - 3 | 393 | 141 | ||
XM_022424289.1 | XP_022279997.1 | 2 - 3 | 666 | 141 | ||
XM_538327.6 | XP_538327.2 | 2 - 3 | 394 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |