Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641988.3 | XP_005642045.1 | 1 | 458 | 141 | |
XM_005641989.3 | XP_005642046.1 | 2 | 358 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |