Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417694.1 | XP_022273402.1 | 2 - 3 | 268 | 146 | |
XM_022417695.1 | XP_022273403.1 | 3 - 4 | 396 | 146 | ||
XM_022417696.1 | XP_022273404.1 | 2 - 3 | 349 | 146 | ||
XM_546238.5 | XP_546238.2 | 2 - 3 | 381 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |