Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_540123.6 | XP_540123.4 | 2 - 3 | 1852 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |