Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022422389.1 | XP_022278097.1 | 3 | 530 | 96 | |
XM_022422390.1 | XP_022278098.1 | 3 | 583 | 96 | ||
XM_022422391.1 | XP_022278099.1 | 3 | 393 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |