Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639867.4 | XP_003639915.1 | 2 - 3 | 476 | 125 | |
XM_005635186.3 | XP_005635243.1 | 2 - 3 | 293 | 125 | ||
XM_014107191.2 | XP_013962666.1 | 3 - 4 | 559 | 125 | ||
XM_862201.4 | XP_867294.2 | 2 - 3 | 293 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |