Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628228.3 | XP_005628285.1 | 3 - 4 | 487 | 158 | |
XM_539280.6 | XP_539280.2 | 3 - 4 | 718 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |