Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426688.1 | XP_022282396.1 | 986 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |