Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022408317.1 | XP_022264025.1 | 3 - 4 | 3367 | 285 | |
XM_534128.6 | XP_534128.3 | 2 - 3 | 3197 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |