Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048112.1 | NP_001041577.1 | 2 - 3 | 787 | 104 | |
XM_014107460.2 | XP_013962935.1 | 3 - 4 | 1450 | 104 | ||
XM_022409564.1 | XP_022265272.1 | 3 - 4 | 1454 | 104 | ||
XM_022409565.1 | XP_022265273.1 | 3 - 4 | 1480 | 104 | ||
XM_022409566.1 | XP_022265274.1 | 3 - 4 | 1367 | 104 | ||
XM_022409567.1 | XP_022265275.1 | 3 - 4 | 1461 | 104 | ||
GenBank mRNA | DQ666280.1 | - | 2 - 3 | 787 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |