Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423299.1 | XP_022279007.1 | 5 | 875 | 131 | |
XM_022423300.1 | XP_022279008.1 | 5 | 898 | 131 | ||
XM_548256.6 | XP_548256.2 | 4 | 676 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |