Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636777.3 | XP_005636834.1 | 3 | 316 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |