Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639263.2 | XP_003639311.1 | 1 | 908 | 81 | |
GenBank mRNA | EF451962.1 | - | 1 | 902 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |