Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097548.1 | NP_001091017.1 | 1 - 2 | 98 | 85 | |
XM_005618263.3 | XP_005618320.1 | 1 - 2 | 163 | 85 | ||
GenBank mRNA | AM422132.1 | - | 1 - 2 | 98 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |