Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBED5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097982.2 | NP_001091451.1 | 3 | 2077 | 142 | |
XM_005633303.3 | XP_005633360.1 | 3 | 2573 | 142 | ||
XM_005633306.3 | XP_005633363.1 | 3 | 2242 | 142 | ||
XM_005633307.3 | XP_005633364.1 | 3 | 2604 | 142 | ||
XM_014122437.2 | XP_013977912.1 | 3 | 2290 | 142 | ||
XM_022407442.1 | XP_022263150.1 | 3 | 2273 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |