Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAL3ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135126.1 | NP_001128598.1 | 1 - 2 | 130 | 75 | |
XM_005636034.3 | XP_005636091.1 | 3 - 4 | 386 | 75 | ||
GenBank mRNA | AB240641.1 | - | 1 - 2 | 130 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |