Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197120.1 | NP_001184049.1 | 1073 | 81 | ||
GenBank mRNA | AB379812.1 | - | 1073 | 81 |